NEBNEXT® ULTRA™ Directional RNA Second Strand Synthesis Module, New England Biolabs
Fournisseur: New England Biolabs (NEB)
Certificats
À propos de cet article
The NEBNext® Ultra™ Directional RNA Second Strand Synthesis Module has been optimized to generate double-stranded cDNA from first-strand cDNA
The dsDNA generated by the NEBNext Ultra Directional RNA Second Strand Module can be subsequently converted to blunt-end DNA fragments using the NEBNext Ultra End Prep Module.
Frequently Bought Together