À propos de cet article
The HiScribe T7 Quick High Yield RNA Synthesis Kit offers robust in vitro RNA transcription for a variety of applications
The master mix format allows for quick reaction setup. DNase I and LiCl are included for DNA template removal and quick RNA purification. In addition, the kit allows for capped RNA or dye labeled RNA synthesis by incorporation of cap analog (ARCA) or dye modified nucleotides
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.