ContactUsLinkComponent
Q5® Site-Directed Mutagenesis Kit (Without Competent Cells), New England Biolabs
Catalogue # CA102877-562
Numéro CAS:  
undefined
Q5® Site-Directed Mutagenesis Kit (Without Competent Cells), New England Biolabs
Catalogue # CA102877-562
Supplier Number:  E0552S
Numéro CAS:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Q5 Site-Directed Mutagenesis Kit (Without Competent Cells)
  • Size:
    10 Reactions
  • Cat. no.:
    CA102877-562
Spécifications
À propos de cet article
The Q5 Site-Directed Mutagenesis Kit (Without Competent Cells) enables rapid, site-specific mutagenesis of double-stranded plasmid DNA in less than 2 hours
The kit utilizes the robust Q5 Hot Start High-Fidelity DNA Polymerase along with custom mutagenic primers to create insertions, deletions and substitutions in a wide variety of plasmids. After PCR, the amplified material is added directly to a unique Kinase-Ligase-DpnI (KLD) enzyme mix for rapid (5 minutes), room temperature circularization and template removal. Transformation into high-efficiency chemically-competent E. coli, not supplied, ensures robust results with plasmids up to at least 20 kb in length.