VWR® Lanced and Unlanced Cylindrical Closures, Stainless Steel
Fournisseur: VWR International
Certificats
À propos de cet article
Non-contaminating, noncorrosive stainless steel cylindrical closures are available in lanced and unlanced formats.
- Prevents dust and microorganism buildup
- Reduce amount of specimen evaporation by 50% compared to cotton plugging
- Withstand repeated autoclaving without loss of resiliency
- Available in plain or lanced formats
- Lanced closures feature three resilient fingers that Adjust to nominal variations in tube O.D.
Top surface indentations allow adequate gas exchange under aerobic and anaerobic conditions.
Frequently Bought Together