- Glove Color:Brown
- Glove size:10
- Ambidextrous/Hand-specific:Hand-specific
- Latex/Latex-free:Latex-free
- Material:Nitrile
- Length:11"
- EN norm:3111
- Cuff style:Safety cuff
- Liner material:Cotton jersey
- Finish:Palm Impregnated
- Cat. no.:CA32890-232
With nitrile-impregnated coating on a jersey liner, these gloves supersede the classic leather general-purpose glove in medium-duty applications.
- Great flexibility and comfort
- Excellent dry grip for quicker, easier handling
- Resist shrinkage in machine washing
- Long-lasting durability ensures economical reuse
These nitrile coated gloves are more flexible, more comfortable, and far more hard-wearing.
Use in the agriculture and viticulture, automotive and transportation, building and construction, chemical, glass, and metal fabrication industries. Uses include assembling metal parts and components; assembly and finishing; gas and electricity supply; general purpose, offshore; handling light castings and metal parts; injection molding; handling cold components; maintenance; mining; operating machinery; and warehouse work.
The high-performance nitrile impregnation offers superior resistance to abrasion and good protection against snags, cuts and puncture. Effectively repels grease, oil and dirt. The soft, flexible jersey lining offers a close, comfortable fit while safely cushioning the hand. Absorbs perspiration, keeping the hand dry and comfortable for longer. Gloves provide excellent dry grip and make handling quicker and easier. Safely machine-washable: will not shrink, stiffen or lose shape. More hygienic and longer lasting, they can safely and economically be reused.