À propos de cet article
In light oil applications, Ansell’s 28-350 gloves provide greater cut, puncture and abrasion resistance than that offered by standard leather-palm gloves.
- Porous coating channels oil away effectively
- Superior flexibility
- Excellent resistance to cuts, punctures, and abrasions
The porous nitrile coating channels oil away from the surface for better grip, while providing superior flexibility, dexterity, and resistance against cuts, punctures, and abrasions.
Use in the appliance manufacturing, automotive, general industry, machining, and metal fabrication industries. Uses include handling abrasive or sharp-edged building materials; handling door panels, hoods and other sharp-edged materials; handling glass or slippery plastics; heavy equipment assembly and cleanup; heavy-duty handling of castings, cores and fabricated metals; servicing and maintaining metalworking equipment; and sheet metal handling and finishing.
The five-piece jersey lining incorporates cut-resistant DuPont™ Kevlar® panels in the high-stress areas for reinforced hand protection with comfort and flexibility.
Spécifications
- Catalog No:
- 80092-700
- 80092-698
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Kevlar®/cotton jersey
- Kevlar®/cotton jersey
- Finish:
- Palm Coated
- Palm Coated
- Cuff style:
- Safety cuff
- Safety cuff
- EN norm:
- 3121
- 3121