To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- No. of channels:1 (measurement)
- Description:Meter only
- Parameters:pH, ORP (mV), DO %, DO mg/L (BOD probe), Partial pressure, Conductivity, Resistivity, Salinity, TDS, Temperature
- Calibration points:DO and Conductivity = 1, pH = 1 to 5
- Data logging capacity:500 data sets in manual mode and 5,000 data sets in automatic mode
- Display:Graphic
- Interfaces:Mini USB; USB-A (PC Connectivity)
- Power supply:Universal power supply
- Temperature compensation:Yes, except ORP
- Cat. no.:10122-580
- Supplier No.:1FD350PY
Spécifications
À propos de cet article
This compact, digital precision meter with integrated printer enables you to carry out pH measurements, ORP measurements, conductivity measurements and dissolved oxygen (D.O.) measurements quickly and reliably.
- One channel input for pH, ORP, DO/BOD, or conductivity
- Data storage - 500 data sets in manual mode and 5,000 data sets in automatic logging mode
- Large, easy to read graphic display
- Intelligent, digital sensors - plug and play sensors
- GLP traceability (sensors store serial # and calibration data)
- Integrated printer enables to document measurements according to GLP requirements
- USB connectivity to manage data
- 3-year warranty
This instrument has one channel to accommodate sensors. Suitable sensors are IDS pH sensors, IDS ORP sensors, IDS conductivity sensors and IDS D.O. sensors. In digital IDS sensors, sensor data are stored that clearly identify the sensor.
The sensor data are automatically taken over by the meter.
Plug and play connectivity with each instrument: IDS senors store their own unique ID with serial number and calibration data. Digital sensor recognition, processing and data transfer. Includes self-stirring BOD, pH, ORP and conductivity sensors. BOD probe uses optical dissolved oxygen technology and includes a guarded sensor tip.
CMC: The CMC function displays continual monitoring of the sensor's measuring range. The range is represented with a bar and calibration points are shown as vertical lines at the cal point. A moving cursor shows the current measured value and indicates if the measurement is within the calibrated range.
Quality Sensor Control: QSC is a system to monitor the condition of the pH electrode. An initial calibration is performed and, afterwards, the sensor status is monitored and graphically displayed over time.