TNE Buffer, Quality Biological
TNE Buffer, Quality Biological
TNE buffer, 500 ml, 7.8 - 8.0
Catalog # 10128-582
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier #:  351-302-101
TNE Buffer, Quality Biological
Catalog # 10128-582
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
TNE Buffer, Quality Biological
TNE Buffer, Quality Biological
TNE buffer, 500 ml, 7.8 - 8.0
CAS Number:  
Price
$112.82
Log in pour la disponibilité
Each (500ml)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
TNE Buffer is often used as a base material to generate lysis buffers.
  • Filtered through a 0.22 µm membrane
  • DNase, RNase, and Protease tested
More specifically, TNE Buffer is useful for: isolation of proteins from yeast cells as a lysis buffer, resuspension of pelleted virus or reporter virus particles and the flurometric determination of DNA content.
Supplier recommendation:
Without correcting the pH, concentrated virus can be used in non-denaturing SDS/PAGE-Western Blot analysis.
If the pH is corrected to inhibit triggering of the viral fusion machinery (for West Nile Virus, this corresponds to pH 7.4), the concentrated virus will remain infectious.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: This product requires special production. Please allow 2 to 4 weeks for delivery.
Caution: Research use only.
Spécifications
Spécifications
  • Désignation:
    TNE buffer
  • Taille:
    500 ml
  • pH:
    7.8 - 8.0
  • Cat. No.:
    10128-582
  • Protease assay:
    Not detectable
  • Test description:
    Endotoxin and mycoplasma tested
  • pH:
    7.8 - 8.0
  • DNase/RNase activity:
    Not detectable
  • Shelf life:
    36 months
  • Sterility:
    Sterile filtered
  • Shipping temperature:
    Ambient