Customized SDS Protein Gel Loading Solution, Quality Biological
This is a special formulation of loading solution for protein gel electrophoresis.
- SDS-PAGE
- Western Blotting
- Analysis of DNA samples with high amounts of DNA binding proteins
- Kinetic experiments
- DNA agarose gel analysis after DNA restriction digestions, ligation, or dephosphorylation
Recommendation
For complex mixtures, use 50 µg total protein per lane.
For purified proteins, use 0.5-5 µg total protein per lane.
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months
: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
: Research use only.
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- Shelf life:36 months
- Shipping temperature:Ambient
Frequently Bought Together