TBS 10X Buffer Solution, pH 7.4, Quality Biological
TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.
- DNase, RNase, and Protease tested
TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications.
Contenst: Tris base, Tris HCl, sodium chloride and Cell Culture Grade Water.
: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
: Research use only.
TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.
- DNase, RNase, and Protease tested
TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications.
Contenst: Tris base, Tris HCl, sodium chloride and Cell Culture Grade Water.
: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
: Research use only.
- Catalog No:
- 10128-546
- 10128-548
- 10128-550
- 10128-552
- 10128-704
- 10128-702
- Test description:
- DNAse, RNAse and protease tested
- DNAse, RNAse and protease tested
- DNAse, RNAse and protease tested
- DNAse, RNAse and protease tested
- DNAse, RNAse and protease tested
- DNAse, RNAse and protease tested
- Sterility:
- Sterile filtered
- Sterile filtered
- Sterile filtered
- Sterile filtered
- Sterile filtered
- Sterile filtered
- Shelf life:
- 36 months
- 36 months
- 36 months
- 36 months
- 36 months
- 36 months
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Product Family Options
- DésignationTaillepH
- : 351-086-101Minority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Cat. No.10128-546Test descriptionDNAse, RNAse and protease testedSterilitySterile filteredShelf life36 monthsShipping temperatureAmbientMinority or Woman-Owned Business EnterpriseSmall Business Enterprise - : 351-086-101CSMinority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Cat. No.10128-548Test descriptionDNAse, RNAse and protease testedSterilitySterile filteredShelf life36 monthsShipping temperatureAmbientMinority or Woman-Owned Business EnterpriseSmall Business Enterprise - : 351-086-131Minority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Cat. No.10128-550Test descriptionDNAse, RNAse and protease testedSterilitySterile filteredShelf life36 monthsShipping temperatureAmbientMinority or Woman-Owned Business EnterpriseSmall Business Enterprise - : 351-086-131CSMinority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Cat. No.10128-552Test descriptionDNAse, RNAse and protease testedSterilitySterile filteredShelf life36 monthsShipping temperatureAmbientMinority or Woman-Owned Business EnterpriseSmall Business Enterprise - : 351-086-491Minority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Cat. No.10128-704Test descriptionDNAse, RNAse and protease testedSterilitySterile filteredShelf life36 monthsShipping temperatureAmbientMinority or Woman-Owned Business EnterpriseSmall Business Enterprise - : 351-086-151Minority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Cat. No.10128-702Test descriptionDNAse, RNAse and protease testedSterilitySterile filteredShelf life36 monthsShipping temperatureAmbientMinority or Woman-Owned Business EnterpriseSmall Business Enterprise