Phase Separation Filter, Grade 480, Ahlstrom
: Ahlstrom
Ahlstrom Grade 480 is a hydrophobic silicone-impregnated paper intended for phase separation.
Frequently Bought Together
Ahlstrom Grade 480 is a hydrophobic silicone-impregnated paper intended for phase separation.