ContactUsLinkComponent
Quartz Microfiber Filter Paper, Grade MK 360, Ahlstrom
Catalog # 10199-564
Supplier:  Ahlstrom
undefined
Quartz Microfiber Filter Paper, Grade MK 360, Ahlstrom
Catalog # 10199-564
Supplier:  Ahlstrom
Supplier Number:  3600-0470
Specifications
  • Size:
    4.7 cm dia.
  • Cat. no.:
    10199-564
  • Supplier No.:
    3600-0470
Specifications
About this item
Micro-quartz filters can be used in the analysis of hot 'stack' gases and other acidic gases (excluding HF).
They contain minimal traces of metals and minerals with each batch trace element certified. Temperature resistant up to 900 °C.