Quartz Extraction Thimbles, Ahlstrom
Catalogue # 10199-580
Fournisseur: Ahlstrom
Spécifications
- Length:123 mm
- Cat. no.:10199-580
- Inner Diameter:43 mm
- Supplier No.:E360-4323
Spécifications
À propos de cet article
Quartz thimbles are used for emission testing in high temperature environments
Wall thickness is approximately 2 mm thick. Temperature resistance of 900 degrees Celsius.