Glass Extraction Thimbles, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Glass thimbles are used to sample dust particles and aerosols from gaseous streams or for Soxhlet extractions requiring solvents too aggressive for cellulose thimbles
Wall thickness is approximately 2 mm thick. Temperature resistance of 500 degrees Celsius.
Frequently Bought Together