To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Dissolved oxygen handheld instrument
- Cat. no.:10065-228
- Supplier No.:6050020
The Pro20 is a handheld dissolved oxygen meter with sensors for wastewater compliance reporting of BOD, CBOD, and DO.
- Rugged, reliable instrument design and user-friendly instrument for dissolved oxygen measurement
- The Pro20 can be used in the lab or the field and can go from one to the other in seconds
- User-replaceable sensors and cables and convenient One Touch Cal feature makes it extremely user friendly
Mil-spec (military spec) connectors, IP-67 waterproof and rubber over molded case, and 1-meter drop tests ensure the instrument remains in your hands to provide years of sampling even in the harshest field conditions
Fast response times with the Polarographic or Galvanic dissolved oxygen sensors allow you to complete your sampling routine quickly. The long life of the Polarographic sensor is notable. The Galvanic sensor enables you to start sampling immediately without a warm-up period. Either sensor you choose for the field will give you accurate, quality data.
Additional features include Internal barometer for highest accuracy. Choose field DO sensors (Polarographic or Galvanic) or lab BOD probe for BOD in wastewater. One Touch Cal calibrates within 3 seconds with the push of a button. Stores 50 data sets. Graphic, backlit display and glow in the dark keypad. Available with 1, 4, 10, 20, 30 or 100-meter cables. View display with polarized sunglasses on. Fastest response time with 95% response in 8 seconds with standard membrane. Multiple languages include English, Spanish, French and German. User replaceable sensors and cables.
The feature set of the Pro20 results in a low total cost of ownership over the life of the product.