À propos de cet article
The YSI MultiLab line includes the 4010-1 (single channel), 4010-2 (dual channel) and 4010-3 (three channel) instruments providing easy to use and calibrate menu-driven operation ideal for the laboratory
- One channel input for pH, ORP, DO/BOD, or conductivity
- 500 data sets in manual mode and 5,000 data sets in automatic logging mode
- Large, easy to read graphic display
- Intelligent, digital sensors (plug and play sensors)
- GLP traceability (sensors store serial # and calibration data)
- USB connectivity to manage data
- Timer: 1 to 999 days
The CMC function displays continual monitoring of the sensor's measuring range. The range is represented with a bar and calibration points are shown as vertical lines at the cal point. A moving cursor shows the current measured value and indicates if the measurement is within the calibrated range. Quality Sensor Control is a system to monitor the condition of the pH electrode. An initial calibration is performed and, afterwards, the sensor status is monitored and graphically displayed over time.
Measure pH, ORP, conductivity or BOD accurately in the laboratory with the YSI 4010-1. This instrument has one channel to accommodate and of the probe choices. The IDS sensors automatically store their unique serial number and calibration data. In addition, they also digitally process the measurement signal. The sensors can be moved from instrument to instrument and maintain their calibration data and transmit this information to the new instrument. Plug and play connectivity with each instrument. IDS sensors store their own unique ID with serial number and calibration data. Digital sensor recognition, processing and data transfer. Includes self-stirring BOD, pH, ORP and conductivity sensors. BOD probe uses optical dissolved oxygen technology and includes a guarded sensor tip.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10066-714
- Temperature compensation:
- Yes; Except ORP
- Display:
- Graphic
- Power supply:
- Universal
- Memory:
- 500 data sets in manual mode and 5,000 data sets in automatic mode
- Interfaces:
- Mini USB; USB-A
- Parameters:
- pH, ORP (mV), DO %, DO mg/L (BOD probe), Partial pressure, Conductivity, Resistivity, Salinity, TDS, Temperature, ISEs
- Calibration:
- DO and Conductivity = 1; pH = 1 to 5; 10 max.