MultiLab IDS Conductivity and Temperature Sensors, Four-Granite Electrodes, YSI
Fournisseur: YSI
Certificats
À propos de cet article
The IDS sensors automatically store their unique serial number and calibration data.
In addition, they also digitally process the measurement signal. The IDS sensors can be moved from instrument to instrument and maintain their calibration data and transmit that information to the new instrument.
Ordering information: Sensors come with a two year warranty.
Spécifications
- Catalog No:
- 10066-690
- 10066-688
- Conductivity range:
- 10 µS/cm to 2,000 mS/cm +0.5% of value
- 10 µS/cm to 2,000 mS/cm +0.5% of value
- Salinity range:
- 0.0 to 70.0 ppt +0.5% of value
- 0.0 to 70.0 ppt +0.5% of value
- TDS range:
- 0 to 1,999 mg/L; 0.0 to 199.9 g/L +0.5% of value
- 0 to 1,999 mg/L; 0.0 to 199.9 g/L +0.5% of value
- Resistivity range:
- 0.5 Ohm cm to 100 kOhm cm +0.5% of value
- 0.5 Ohm cm to 100 kOhm cm +0.5% of value
- Cell constant:
- 0.475 cm +/-1.5%
- 0.475 cm +/-1.5%
- Temperature accuracy:
- +/-0.2 °C
- +/-0.2 °C