The YSI MultiLab line includes the 4010-1 (single channel), 4010-2 (dual channel) and 4010-3 (three channel) instruments providing easy to use and calibrate menu-driven operation ideal for the laboratory
- One channel input for pH, ORP, DO/BOD, or conductivity
- 500 data sets in manual mode and 5,000 data sets in automatic logging mode
- Large, easy to read graphic display
- Intelligent, digital sensors (plug and play sensors)
- GLP traceability (sensors store serial # and calibration data)
- USB connectivity to manage data
- Includes self-stirring BOD, pH, ORP and conductivity sensors
Measure pH, ORP, conductivity or BOD accurately in the laboratory with the YSI 4010-1. This instrument has one channel to accommodate and of the probe choices. The IDS sensors automatically store their unique serial number and calibration data. In addition, they also digitally process the measurement signal. The sensors can be moved from instrument to instrument and maintain their calibration data and transmit this information to the new instrument. Plug and play connectivity with each instrument
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 10066-708
- pH calibration:
- 1 - 5
- Conductivity calibration:
- 1
- DO calibration:
- 1
- Temperature compensation:
- Yes; Except ORP
- Display:
- Large backlit graphic display
- Power supply:
- Universal
- Memory:
- 500 data sets in manual mode and 5,000 data sets in automatic mode
- Interfaces:
- Mini USB; USB-A
- Parameters:
- pH, ORP (mV), DO %, DO mg/L (BOD probe), Partial Pressure, Conductivity, Resistivity, Salinity, TDS, Temperature