HT5 GC Capillary Columns, SGE
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
- Ultra high temperature columns
- Unique phases
- Ideal for simulated distillation applications (petroleum industry)
- Bonded and cross-linked
- Able to be solvent rinsed
These columns can be used in simulated distillation, general hydrocarbon profiles, pesticides/herbicides and GC/MS applications.
: RoHS compliant per Directive 2011/65/EU
: The 5% phenyl polycarborane-siloxane columns are a suitable replacement for MXT-1 Sim Dist, HT-Sim, DistCB and MXT-500.
- Catalog No:
- 14226-002
- 60361-774
- 14226-006
- 60361-754
- 60361-776
- 60361-766
- 60361-740
- 60361-768
- 60361-742
- 60361-756
- 60361-758
- 14226-004
- 60361-736
- Description:
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- GC Capillary Column, (5% Phenyl Polycarborane-siloxane)
- Temperature limit:
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
- 10 to 380/400 °C
Frequently Bought Together