Automatic Digital Refractometers, ATAGO® | Avantor
Automatic Digital Refractometers, ATAGO®
Supplier:   Atago
Automatic Digital Refractometers, ATAGO®
Supplier:  Atago
Certificates
Automatic Digital Refractometers, ATAGO®
Automatic Digital Refractometers, ATAGO®
4 Options Available Below
Product Details & Documents
These digital refractometers offer automatic and accurate measurement of the refractive index and Brix scale.
Model RX-5000i-plus and RX-5000i are designed for the measurement of wide range of samples including emulsions. It has a measurement temperature of 5 to 75 °C (41 to 167 °F).
All models have a printer output, as well as a USB for connection to a computer.
Ordering information: Supplied with a one-year manufacturer’s warranty. A calibration certificate is available for each unit. Contact your Avantor representative for information.
Product Family Options
Informations sur le produit
  • Modèle
    Electrical
    Résolution
    L×W×H
    Accuracy
    Measurement Range
    Weight
    Availability
    Price
  • RX-5000i
    90 VA
    0.00001 nD 0.01% Brix
    37 x 26 x 14 cm (149/16 x 101/4 x 51/2")
    ±0.00004 nD ±0.03% brix
    1.32422–1.58000 nD 0.00–100.00% Brix
    6.6 kg (14.55 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $43,845.38
    $43,845.38
    Each
       $43,845.38
    Supplier Number: 3276
  • RX-5000i-Plus
    90 VA
    0.00001 nD 0.005% Brix
    37 x 26 x 14 cm (149/16 x 101/4 x 51/2")
    ±0.00002 nD ±0.010% brix
    1.32422–1.58000 nD 0.00–100.00% Brix
    6.6 kg (14.55 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $55,485.18
    $55,485.18
    Each
       $55,485.18
    Supplier Number: 3275
Digital Printers
Informations sur le produit
  • Modèle
    Electrical
    Résolution
    L×W×H
    Accuracy
    Measurement Range
    Weight
    Availability
    Price
  • DP-63
    13 VA
    17 x 16 x 7 cm (7 x 6 x 3")
    0.58 kg (1 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,248.50
    $5,248.50
    Each
       $5,248.50
    Supplier Number: 3118-DP-63
  • DP-AD
    7 VA
    11 x 18 x 9 cm (45/16 x 7 x 31/2")
    0.47 kg (1 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,664.36
    $5,664.36
    Each
       $5,664.36
    Supplier Number: 3123