Polycool PG -20 Fluid, PolyScience | Avantor
Polycool PG -20 Fluid, PolyScience
Propylene glycol, 3.8 l (1 gal.), –20...+100 °C (–4...+212 °F), when mixed 50/50 with distilled water
Catalog # 71002-506
Supplier:   Polyscience
Fournisseur #:  060320
Polycool PG -20 Fluid, PolyScience
Catalog # 71002-506
Supplier:  Polyscience
CAS Number:  
Polycool PG -20 Fluid, PolyScience
Polycool PG -20 Fluid, PolyScience
Propylene glycol, 3.8 l (1 gal.), –20...+100 °C (–4...+212 °F), when mixed 50/50 with distilled water
CAS Number:  
Price
$457.69
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each (1US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Propylene glycol is a safer alternative to ethylene glycol.
For use with VWR® Refrigerated Circulating Baths, VWR® Benchtop Chillers and VWR® Recirculating Chillers.
Lowers freezing point of water to allow circulation at below freezing temperatures. Mix with distilled water for broadest temperature range.
Spécifications
Spécifications
  • Désignation:
    Propylene glycol, 3.8 l (1 gal.)
  • Temperature Range:
    –20...+100 °C (–4...+212 °F), when mixed 50/50 with distilled water