Polycool EG -25 Fluid, PolyScience
Ethylene glycol, 3.8 l (1 gal.), –25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled water
Catalog # 71002-504
Supplier:   Polyscience
 :  060340
Polycool EG -25 Fluid, PolyScience
Catalog # 71002-504
Supplier:  Polyscience
CAS Number:  
Polycool EG -25 Fluid, PolyScience
Polycool EG -25 Fluid, PolyScience
Ethylene glycol, 3.8 l (1 gal.), –25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled water
CAS Number:  
Price
$253.34
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each (1US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A common circulator anti-freeze fluid.
For use with VWR® Refrigerated Circulating Baths, VWR® Benchtop Chillers and VWR® Recirculating Chillers.
Lowers the freezing point of water to allow circulation at below freezing temperatures. High toxicity. Recommend mixing with distilled water for broadest temperature range.
Spécifications
Spécifications
  • Désignation:
    Ethylene glycol, 3.8 l (1 gal.)
  • Temperature Range:
    –25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled water