Specimen Containers with Temperature Strip for Drug Testing, Starplex Scientific
Supplier: Starplex
Certificates
About this item
These drug testing containers are available in regular and wide mouth 90 mL capacities
Containers have an orange or white o-ring cap with applied temperature strip and tamper evident patient identification labels. The temperature strip measures a range from 32°C to 38°C (90° F to 100°F).
Ordering information: Containers are packaged 400 per case.
Frequently Bought Together