ContactUsLinkComponent
Electrical Lockout Pouch, Brady Worldwide®
Electrical Lockout Pouch, Brady Worldwide®
Catalog # CA12222-708
Supplier:  Avantor
undefined
Electrical Lockout Pouch, Brady Worldwide®
Catalog # CA12222-708
Supplier:  Avantor
Specifications
  • Description:
    Electrical lockout pouch
  • Cat. no.:
    CA12222-708
  • W×D×H:
    7.5×2×4.75"
  • Supplier No.:
    65291
Specifications
About this item
Lightweight, portable pouch contains lockout devices to secure electrical equipment.
  • Metal loops allow for hanging on a belt
Pouch contents include; one 2" blue padlock, one red labeled lockout hasp, one universal multi pole breaker lockout, one 277 Volt 'No Hole' circuit breaker lockout, one small red plug lockout, and two heavy-duty lockout tags.