Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific | Avantor
Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific
Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific
Supplier:  Starplex
Certificates
Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific
Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific
1 Options Available Below
Product Details & Documents
This premium quality line of containers consists of 60 mL, 90 mL, wide 90 mL and 120 mL sizes for leakproof transportation
Each polypropylene container is lot numbered and has clear graduations. The wide 90 mL and 120 mL containers feature a QuickTurn™ cap closure for easy application. Sterile containers have an orange cap and tamper tab patient label. The patient identification labels meet current JCAHO standards. Non-sterile containers have a yellow cap and a non-tabbed patient label. The standard 90 mL container is also available in an opaque polypropylene for discrete transport. A space saver dispenser box of 20, 90 mL containers is convenient for laboratories and Doctor's offices. Ideal for Operating Room use, both the 90 mL and 120 mL containers are packaged in a sterile, clear plastic peel pouch.
Ordering information: Leakbuster containers are packaged 100 containers per case.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Availability
    Price
  • 120 ml
    Leakbuster™ Containers
    Discontinued
    Supplier Number: B1202-1W-OR
  • 120 ml
    Leakbuster™ Containers
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $198.00
    $198.00
    Case of 100
       $198.00
    Supplier Number: B1202-1W-OR