Cable Attachments, Brady Worldwide® | Avantor
Cable Attachments, Brady Worldwide®
Supplier:   Brady Worldwide
Cable Attachments, Brady Worldwide®
Supplier:  Brady Worldwide
Certificates
Cable Attachments, Brady Worldwide®
Cable Attachments, Brady Worldwide®
2 Options Available Below
Product Details & Documents
Cable attachments are designed to lock out gate valves
Cable attachments are available in two formats. Choose between a 1/8" nonconductive nylon cable or a rustproof sheathed metal cable.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Longueur
    Availability
    Price
  • Nylon cable attachment
    Red
    2.4 m (8 ft.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.89
    $34.89
    Each
       $34.89
    La quantité minimale de commande est de 1
    Supplier Number: 50932
  • Sheathed cable attachment
    Red
    2.4 m (8 ft.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.58
    $35.58
    Each
       $35.58
    La quantité minimale de commande est de 1
    Supplier Number: 51395