Lockout Satchel, Brady Worldwide®
CA89007-248
- Color:Black on Red
- Description:Lockout satchel
- Cat. no.:CA89007-248
- W×D×H:381×102×335 mm
- Weight:102 mm
- Supplier No.:51173
The new lockout satchel is made of 1000 Denier red cordura nylon with a 99 cm (39") should strap.
: Satchel includes latchable front pocket and two side mesh holders.