SP Bel-Art Stainless Steel Clamping Thermometer Holder, Bel-Art Products, a part of SP
Catalog # 10204-914
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Thermometer Holder
- Height:2½"
- Length:3¼"
- Cat. no.:10204-914
- Width:2⅛"
- Supplier No.:B61400-7300
Specifications
About this item
With five hole sizes ranging from 3.5 mm to 14.5 mm, this holder accommodates various thermometers including liquid in glass, electronic with probe, and bimetal with probe.
- Stainless steel construction
- Durable polyurethane foam keeps thermometers securely in place
- Rubber bumpers prevent scratching
The stainless steel, spring-loaded clip attaches securely to laboratory containers, shelves up to 3/8" thick, and more. Rubber bumpers on the clip prevent scratching. Durable polyurethane foam provides a secure grip to keep thermometers in place.