Safe-T-Grip® Fuse Puller, Brady | Avantor
Safe-T-Grip® Fuse Puller, Brady
Supplier:   Avantor
Safe-T-Grip® Fuse Puller, Brady
Supplier:  Avantor
Certificates
Safe-T-Grip® Fuse Puller, Brady
Safe-T-Grip® Fuse Puller, Brady
2 Options Available Below
Product Details & Documents
Remove or replace cartridge fuses with ease by using the Safe-T-Grip® fuse pullers.
Notched handles ensure a no-slip grip. Amperage ranges are clearly marked and both types exceed MIL specifications. These yellow fuse pullers are designed for use with 0 to 100 A, 250 and 600 V fuses.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Small Safe-T-Grip® Fuse Puller
    12.7 cm (5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.67
    $15.67
    Each
       $15.67
  • Large Safe-T-Grip® Fuse Puller
    19 cm (7.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $23.94
    $23.94
    Each
       $23.94