Fuse Puller With Testlite, Brady
Fuse Puller With Testlite, 20.3 cm (8")
Catalog # CA12222-730
Supplier: Avantor
Jump to:
Product Details & Documents
Handy pocket-sized yellow-colored tool provides maximum safety when pulling cartridge fuses or checking electrical connections.
One end is a fuse puller and the opposite end features test prods that fit both parallel and tandem slot receptacles. Use on fuses up to 100A/250V and 60A/600V.
Specifications
- Désignation:Fuse Puller With Testlite
- Longueur:20.3 cm (8")