NOVA-TECH 10 Cobalt Blue, Sterile, Poly-Cellulose Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
Certificates
About this item
NOVA-TECH 10 wipes are made of a synergistic blend of virgin polyester and cellulose that is hydroentangled into a uniform, super-tough fabric. These wipes are converted in a class 100 cleanroom.
- Validated to SAL (Sterility Assurance Level) 10-6 in accordance with ANSI/AAMI/ISO 11137:2006 guidelines
- Cobalt-60 gamma irradiated
- ISO Class 5
- Meets and exceeds USP 797 and USP 800 requirements for critical contamination control
- Each packaged identified with lot number for traceability
- Each case includes Certificate of Sterility
Frequently Bought Together