VWR® A Balances
About this item
VWR® A-Series balances incorporate state-of-the-art features for advanced weighing applications and documentation.
- State-of-the-art features for advanced weighing and documentation
- Internal calibration
- Multiple weigh modes
- 2×RS-232, USB (A and B), and Wi-Fi communication interfaces
- Easy-to-read, adjustable, backlit display
- Tool-free assembly and diassembly of the 3-door draft shield on analytical models
- 5 year warranty
The VWR® A-Series balances are the answer for organizations demanding accuracy and performance at an economical price. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-Series provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100,000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its unsurpassed 5-year warranty, the VWR® A-Series balance will provide reliable results for years to come.
14 Options Available Below
Product Details & Documents
VWR® A-Series balances incorporate state-of-the-art features for advanced weighing applications and documentation.
- State-of-the-art features for advanced weighing and documentation
- Internal calibration
- Multiple weigh modes
- 2×RS-232, USB (A and B), and Wi-Fi communication interfaces
- Easy-to-read, adjustable, backlit display
- Tool-free assembly and diassembly of the 3-door draft shield on analytical models
- 5 year warranty
The VWR® A-Series balances are the answer for organizations demanding accuracy and performance at an economical price. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-Series provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100,000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its unsurpassed 5-year warranty, the VWR® A-Series balance will provide reliable results for years to come.
Specifications for Product Family
Specifications
- Catalog No:
- 76446-812
- 76446-810
- 76446-814
- 76446-816
- 76446-818
- 76446-820
- 10204-966
- 76300-142
- 76300-144
- 76300-146
- 10204-968
- 10204-970
- 76446-822
- 76300-152
- Modèle:
- VWR-205AC
- VWR-105AC
- VWR-160AC
- VWR-220AC
- VWR-310AC
- VWR-520AC
- VWR-363AC
- VWR-603AC
- VWR-1003AC
- VWR-2102AC
- VWR-4502AC
- VWR-6002AC
- VWR-8102AC
- VWR-10102AC
- Communication interface:
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB-A, USB-B, and Wi-Fi
Specifications
Product Family Options
Analytical with Draft Shield
Product Information
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- VWR-205AC82/220 g0.05 / 0.2 mgØ 90 mm0.01 / 0.1 mg2 s0.02 mg (at 5% load)Log in for availabilityEach$7,857.67Specifications:
More
Specifications
Cat. No.76446-812ModèleVWR-205ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - Specifications:
More
Specifications
Cat. No.76446-810ModèleVWR-105ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - Specifications:
More
Specifications
Cat. No.76446-814ModèleVWR-160ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - Specifications:
More
Specifications
Cat. No.76446-816ModèleVWR-220ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - Specifications:
More
Specifications
Cat. No.76446-818ModèleVWR-310ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - Specifications:
More
Specifications
Cat. No.76446-820ModèleVWR-520ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi
Precision with Short Open-Air Draft Shield
Product Information
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- VWR-363AC360 g0.002 g128×128 mm0.001 g2 s0.0005 g (at 5% load)Log in for availabilityEach$2,207.16Specifications:
More
Specifications
Cat. No.10204-966ModèleVWR-363ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - VWR-603AC600 g0.003 g128×128 mm0.001 g2 s0.0005 g (at 5% load)Log in for availabilityEach$3,738.25Specifications:
More
Specifications
Cat. No.76300-142ModèleVWR-603ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - VWR-1003AC1000 g0.003 g128×128 mm0.001 g2 s0.0005 g (at 5% load)Log in for availabilityEach$4,575.51Specifications:
More
Specifications
Cat. No.76300-144ModèleVWR-1003ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi
Precision, No Draft Shield
Product Information
- ModèleWeighing CapacityLinearityPlateau de pesée/taille de la plateformeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- VWR-2102AC2100 g0.02 g195×195 mm0.01 g1.5 s0.005 g (at 5% load)Log in for availabilityEach$2,189.20Specifications:
More
Specifications
Cat. No.76300-146ModèleVWR-2102ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - VWR-4502AC4500 g0.02 g195×195 mm0.01 g1.5 s0.005 g (at 5% load)Log in for availabilityEach$2,208.74Specifications:
More
Specifications
Cat. No.10204-968ModèleVWR-4502ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - VWR-6002AC6000 g0.03 g195×195 mm0.01 g1.5 s0.005 g (at 5% load)Log in for availabilityEach$3,412.47Specifications:
More
Specifications
Cat. No.10204-970ModèleVWR-6002ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - VWR-8102AC8100 g0.03 g195×195 mm0.01 g1.5 s0.005 g (at 5% load)Log in for availabilityEach$4,464.74Specifications:
More
Specifications
Cat. No.76446-822ModèleVWR-8102ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi - VWR-10102AC10100 g0.03 g195×195 mm0.01 g1.5 s0.005 g (at 5% load)Log in for availabilityEach$5,204.25Specifications:
More
Specifications
Cat. No.76300-152ModèleVWR-10102ACCommunication interfaceRS232 (2), USB-A, USB-B, and Wi-Fi
Accessories
Product Information
- Catalog #AvailabilityPrice
- Density kit, A-series and T-series balances, analytical (0.1 mg) and semi micro (0.01 mg) model onlyDensity kit, A-series and T-series balances, analytical (0.1 mg) and semi micro (0.01 mg) model only10799-182Log in for availabilityEach - $1,735.64