The ODO200 is designed for quick, accurate results in an economical platform. The ability to measure DO optically along with temperature, in a simple, compact handheld allows the instrument to be used across multiple application sampling strategies in the field.
- Automatic temperature compensation
- 1-year instrument, cable and sensor cap warranty
- 1-, 4- and 10-meter durable field cables available
- Manual input for salinity and pressure compensation: 0.0 to 40.0 ppt (manual input)/ 500 to 1125 millibar (manual input)
- Low battery indicator with 100 hour battery life
- Replaceable optical sensor cap (12-18 month life)
- Auto shutoff function after 30 minutes of inactivity
- 50 data set reviewable memory
- IP67 waterproof rating
With a one-year instrument, probe and sensor cap warranty, waterproof case and ease-of-use, the EcoSense ODO200 will fit your needs. Optical benefits include no stirring requirement; no warm-up time; 1-, 4- and 10-meter durable field cables available; no electrolyte solutions; no electrode maintenance; no membranes; no interferences from gases such as hydrogen sulfide.
The ODO200 simultaneously measures dissolved oxygen, using optical technology, and temperature. Accurate, economical, handheld dissolved oxygen measurements using optical technology.
- Catalog No:
- 10146-170
- 10146-166
- 10146-168
- 10146-182
- DO range:
- 0 to 20 mg/L
- 0 to 20 mg/L
- 0 to 20 mg/L
- 0 to 20 mg/L
- DO resolution:
- 0.01 mg/L
1 or 0.1% air saturation - 0.01 mg/L
1 or 0.1% air saturation - 0.01 mg/L
1 or 0.1% air saturation - 0.01 mg/L
1 or 0.1% air saturation
- DO accuracy:
- ±1.5% of reading or ±0.15 mg/L; whichever is greater
±1.5% of reading or ±1.5%; whichever is greater (% air) - ±1.5% of reading or ±0.15 mg/L; whichever is greater
±1.5% of reading or ±1.5%; whichever is greater (% air) - ±1.5% of reading or ±0.15 mg/L; whichever is greater
±1.5% of reading or ±1.5%; whichever is greater (% air) - ±1.5% of reading or ±0.15 mg/L; whichever is greater
±1.5% of reading or ±1.5%; whichever is greater (% air)
- DO saturation:
- 0 to 200%
- 0 to 200%
- 0 to 200%
- 0 to 200%
- Temperature range:
- 0 to +50°C (32 to 122°F)
- 0 to +50°C (32 to 122°F)
- 0 to +50°C (32 to 122°F)
- 0 to +50°C (32 to 122°F)
- Temperature resolution:
- 0.1°C (0.2°F)
- 0.1°C (0.2°F)
- 0.1°C (0.2°F)
- 0.1°C (0.2°F)
- Temperature accuracy:
- ±0.3 °C (±0.6 °F)
- ±0.3 °C (±0.6 °F)
- ±0.3 °C (±0.6 °F)
- ±0.3 °C (±0.6 °F)
- Battery life:
- 100 hours in manual mode; low battery indicator
- 100 hours in manual mode; low battery indicator
- 100 hours in manual mode; low battery indicator
- 100 hours in manual mode; low battery indicator
- Memory:
- 50 data set reviewable memory
- 50 data set reviewable memory
- 50 data set reviewable memory
- 50 data set reviewable memory
- Dimensions:
- 78W x 184L mm (3.1 x 7.25")
- 78W x 184L mm (3.1 x 7.25")
- 78W x 184L mm (3.1 x 7.25")
- 78W x 184L mm (3.1 x 7.25")
- Weight:
- 272 g (0.6 lb.) with battery
- 272 g (0.6 lb.) with battery
- 272 g (0.6 lb.) with battery
- 272 g (0.6 lb.) with battery
- Operating temperature range:
- 0.0 to 50.0°C (32.0 to 122.0°F)
- 0.0 to 50.0°C (32.0 to 122.0°F)
- 0.0 to 50.0°C (32.0 to 122.0°F)
- 0.0 to 50.0°C (32.0 to 122.0°F)
- Operating humidity range:
- up to 95%, RH
- up to 95%, RH
- up to 95%, RH
- up to 95%, RH