Human CD14+ Monocytes (hMoCD14+-PB), PromoCell
PromoCell Human CD14+ Monocytes are isolated from fresh peripheral blood. After isolation of the ultra-pure mononuclear cell fraction by proprietary methods, the CD14+ Monocytes are isolated by positive selection with > 95% purity.
In addition, hMoCD14+-PB are available which are pre-screened for their differentiation capacity into dendritic cells.
This product can be used in the following applications. Dendritic Cell Research, (Auto-)Immunity, Infection biology
: Human CD14+ cells can be orderd as cryopreserved cells. Additional cell pellets stored in RNAlater® are available.
Frequently Bought Together