- Cell Format:Cryopreserved
- Description:From Single Donor immediately after isolation, the freshly prepared CD34+ Progenitor Cells are cryopreserved
- Size:100000 Cryopreserved Cells
- Cat. no.:10175-190
- Supplier No.:C-12921
PromoCell offers CD34+ Progenitor Cells from the cord blood of healthy donors. The CD34+ Progenitor Cells contain two main cellular subpopulations, hematopoietic and endothelial progenitor cells. Therefore, CD34+ Progenitor Cells are suitable for a series of studies, e.g. directed differentiation into more committed types of blood cells and endothelial lineages.
CD34 is a glycosylated transmembrane protein and represents a well-known marker for primitive blood- and bone marrow-derived progenitor cells, especially for hematopoietic and endothelial stem cells
This product can be used in the following applications. Stem cell research, Transplantation basic research, Hematological and immunological studies, Vascular and angiogenesis research, Oncology research
Although the biological functions of CD34 are largely unknown, recent data suggest that CD34 is involved in maintenance of the progenitor cells in a phenotypically undifferentiated state.