Human Pericytes from Placenta (hPC-PL), PromoCell
Fournisseur: Promocell
Certificats
À propos de cet article
Pericytes are multipotent mesenchymal-like cells found in association with small blood vessel walls. They are important for angiogenesis, the structural integrity of the microvasculature, and blood flow regulation. However, they can also develop into malignant tumors called hemangiopericytomas.
To date, pericytes have demonstrated the ability to differentiate into adipocytes, osteoblasts, fibroblasts, smooth muscle cells, and phagocytes (macrophages).
This product can be used in the following applications. Angiogenesis research, Regenerative medicine research, Oncology
Ordering information: Human Pericytes from human placenta can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Frequently Bought Together