ContactUsLinkComponent
Macrophage Detachment Solution DXF, PromoCell
Macrophage Detachment Solution DXF, PromoCell
Catalog # 10175-308
Supplier:  Promocell
undefined
Macrophage Detachment Solution DXF, PromoCell
Catalog # 10175-308
Supplier:  Promocell
Supplier Number:  C-41330
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Macrophage Detachment Solution DXF
  • Size:
    250 ml
  • Cat. no.:
    10175-308
  • Supplier No.:
    C-41330
Specifications
About this item
The Macrophage Detachment Solution DXF is a non-enzymatic reagent especially optimized for the gentle detachment of strongly adherent macrophages.
  • Animal component-free
Macrophages are strongly adherent and are not effectively dislodged from culture surfaces by standard enzymatic cell dissociation reagents. Indeed, these cells are highly sensitive to improper dissociation procedures. It was especially designed for the gentle release of adherent macrophages and guarantees the best possible cell viability, even after prolonged exposure times. Unlike enzyme-based solutions the PromoCell Macrophage Detachment Solution DXF does not alter cell surface proteins and neutralization is not necessary.