Monocyte Attachment Medium, PromoCell
250 mL
Catalog # 10172-398
Supplier:   Promocell
 :  C-28051
Monocyte Attachment Medium, PromoCell
Catalog # 10172-398
Supplier:  Promocell
Monocyte Attachment Medium, PromoCell
Monocyte Attachment Medium, PromoCell
250 mL
Price
$318.70
Log in pour la disponibilité
Each (250ml)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The Monocyte Attachment Medium allows for the efficient adherence selection of Monocytes from freshly isolated Mononuclear Cells (MNC) while maintaining optimal cell health
Recommended for Human Mononuclear Cells, freshly isolated
Thus, the time-consuming and costly immunomagnetic purification of monocytes can be skipped. With properly performed washing steps (see Instruction Manual) large numbers of 80-90% pure monocytes can be obtained with the PromoCell Monocyte Attachment Medium. Typical applications are generation of dendritic cells / macrophages as well as phagocytosis assays.
 : The Medium is ready-to-use and does not need further supplementation.
Spécifications
  • Taille:
    250 mL