Ear Plug Tray, Brady®
Earplug tray, acrylic, Clear, 10.6×10.6×10.6"
Catalog # CA82004-274
Supplier: Brady Worldwide
: PD228E
Jump to:
Product Details & Documents
Clear acrylic ear plug dispenser tray holds a standard box of ear plugs and the front section catches spills and protects the corrugated dispenser
Use on table top or wall mount.
Spécifications
- Color:Clear
- Désignation:Earplug tray, acrylic
- W×D×H:10.6×10.6×10.6"
- Cat. No.:CA82004-274
- Dispenser type:Bench top/wall-mountable
- Material:Acrylic
- No. of compartments:1
- PPE type:Earplugs