ContactUsLinkComponent
Miscellaneous PPE Dispenser, Brady®
Miscellaneous PPE Dispenser, Brady®
  CA87001-940
undefined
Miscellaneous PPE Dispenser, Brady®
  CA87001-940
 :  MPPE
  • Dispenser type:
    Bench top/wall-mountable
  • Material:
    Acrylic
  • No. of compartments:
    1
  • PPE type:
    Multi-use
  • Color:
    Clear
  • Description:
    Multi-use PPE dispenser, acrylic
  • Cat. no.:
    CA87001-940
  • W×D×H:
    9.5×11.5×9.5"
  • Supplier No.:
    MPPE
Clear acrylic dispenser is as versatile as you need it to be and comes with a dust cover lid and a foam bottom
Dispenser can be wall or table mounted.