Human Small Airway Epithelial Cells (HSAEpC), PromoCell
Fournisseur: Promocell
Certificats
À propos de cet article
Primary Human Small Airway Epithelial Cells (HSAEpC) are isolated from the distal portion of the human respiratory tract in the 1 mm bronchiole area. They stain positive for cytokeratin.
- Cytokeratin positive
- Routinely morphology checked
- Cryopreserved at second passage
The distal respiratory tract mainly consists of pulmonary alveoli, which are spherical outcroppings of the respiratory bronchioles and are the primary sites of gas exchange with the blood
This product can be used in the following applications. Drug uptake studies, Toxicological research, Wound healing research, Oncology, Cell differentiation studies, Respiratory disease research.
HSAEpC are qualified for functional studies to investigate disorders such as asthma or pulmonary inflammation.
Ordering information: Human Small Airway Epithelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Frequently Bought Together