Fibroblast Growth Medium for Juvenile Cells, PromoCell
Supplier: Promocell
Certificates
About this item
The PromoCell Fibroblast Growth Medium is a serum-free medium optimized for the cultivation of neonatal and juvenile fibroblasts
Recommended for Normal Human Dermal Fibroblasts (NHDF), juvenile foreskin
The medium is optimized for primary human cells but can also be used for rat, porcine, and bovine fibroblasts as well as fibroblast cell lines
Ordering information: The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.
Frequently Bought Together