Preadipocyte/Adipocyte Medium, PromoCell
Supplier: Promocell
Certificates
About this item
The PromoCell Preadipocyte Growth Medium is a low-serum (5% V/V) medium optimized for the expansion of human preadipocytes
Recommended for Human White Preadipocytes (HWP), Human Adipocytes
Ordering information: PromoCell Preadipocyte/Adipocyte Media are available as Medium (Ready-touse) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as SupplementMix and SupplementPacks can also be purchased separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together