3" Sonic Sifter Sieves, Advantech
À propos de cet article
These 3" diameter "see-through" acrylic framed sieves are made of stainless steel wire cloth and specifically designed to be used with the Sonic Sifter Separator.
- Ideal for particles ranging from 20 µm to 5.6 mm
Centerline® Sieve Inspection or Calibration Certification reports provide detailed statistical data that is linked to each serialized sieve and becomes an integral part of your Quality Assurance Program.
Certifications: Conforms to ASTM E11 specification
Ordering information: Certifications are available as accessories.
33 Options Available Below
Product Details & Documents
These 3" diameter "see-through" acrylic framed sieves are made of stainless steel wire cloth and specifically designed to be used with the Sonic Sifter Separator.
- Ideal for particles ranging from 20 µm to 5.6 mm
Centerline® Sieve Inspection or Calibration Certification reports provide detailed statistical data that is linked to each serialized sieve and becomes an integral part of your Quality Assurance Program.
Certifications: Conforms to ASTM E11 specification
Ordering information: Certifications are available as accessories.
Specifications for Product Family
Spécifications
- Catalog No:
- 57333-752
- 57333-754
- 57333-756
- 57333-758
- 57333-760
- 57333-762
- 57333-764
- 57333-766
- 57333-768
- 57333-770
- 57333-772
- 57334-000
- 57333-774
- 57334-002
- 57333-776
- 57334-004
- 57334-006
- 57334-008
- 57334-010
- 57334-012
- 57334-014
- 57334-016
- 57334-018
- 57334-020
- 57334-022
- 57334-024
- 57334-026
- 57334-028
- 57334-030
- 57333-778
- 57333-780
- 57333-782
- 57333-785
- Int.Ø:
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- Ext.Ø:
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
Spécifications
Product Family Options
Informations sur le produit
- HauteurMesh SizeUSA SeriesAvailabilityPrice
- Supplier Number: L3-S3.5Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-752Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S4Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-754Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S5Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-756Int.ØExt.ØSmall Business Enterprise - Supplier Number: L3-S6Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-758Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S7Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-760Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S8Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-762Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S10Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-764Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S12Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-766Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S14Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-768Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S16Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-770Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S18Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-772Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S20Spécifications:
More
Specifications
Cat. No.57334-000Int.Ø3"Ext.Ø3 1/2" - Supplier Number: L3-S25Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-774Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S30Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-002Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S35Spécifications:
More
Specifications
Cat. No.57333-776Int.Ø3"Ext.Ø3 1/2" - Supplier Number: L3-S40Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-004Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S45Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-006Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S50Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-008Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S60Spécifications:
More
Specifications
Cat. No.57334-010Int.Ø3"Ext.Ø3 1/2" - Supplier Number: L3-S70Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-012Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S80Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-014Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S100Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-016Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S120Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-018Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S140Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-020Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S170Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-022Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S200Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-024Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S230Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-026Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S270Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-028Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S325Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57334-030Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S400Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-778Int.Ø3"Ext.Ø3 1/2"Small Business Enterprise - Supplier Number: L3-S450Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-780Int.ØExt.ØSmall Business Enterprise - Supplier Number: L3-S500Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-782Int.ØExt.ØSmall Business Enterprise - 20 µm (0.0008")#635Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,130.27$1,130.27Prix promotionnel pour les nouveaux clientsEach$1,130.27Supplier Number: L3-S635Spécifications:Small Business Enterprise
More
Specifications
Cat. No.57333-785Int.ØExt.ØSmall Business Enterprise