Close-it®, Tapes, Bürkle
À propos de cet article
With its extremely strong adhesive strength, close-it® tapes adhere to a range of different surfaces, where the glue effect of normal adhesive tape quickly reaches its maximum capacity.
- Customisable
- White with tear-off lines
- Available in the classic close-it® design, as well as close-it® tape food, with a food-compatible adhesive
- Can be labelled with permanent ink and provides gas-proof seal
Standard close-it® and close-it® food control seals are also available on a continuous roll. This offers the flexibility to provide a small length of tape to seal a small hole or to mark a specific point. Alternatively longer pieces can be used to repair a long tear quickly, reliably and easily.
2 Options Available Below
Product Details & Documents
With its extremely strong adhesive strength, close-it® tapes adhere to a range of different surfaces, where the glue effect of normal adhesive tape quickly reaches its maximum capacity.
- Customisable
- White with tear-off lines
- Available in the classic close-it® design, as well as close-it® tape food, with a food-compatible adhesive
- Can be labelled with permanent ink and provides gas-proof seal
Standard close-it® and close-it® food control seals are also available on a continuous roll. This offers the flexibility to provide a small length of tape to seal a small hole or to mark a specific point. Alternatively longer pieces can be used to repair a long tear quickly, reliably and easily.
Product Family Options
Standard
Informations sur le produit
- DésignationLongueurLargeurAvailabilityPrice
- Supplier Number: 5303-0017
Food
Informations sur le produit
- DésignationLongueurLargeurAvailabilityPrice
- Supplier Number: 5303-2019