Valve Brushes, Micronova
Supplier: Micronova
Certificates
About this item
The valve brush is a large, oval-shaped, polyester bristle brush with a plastic core and handle, designed for equipment cleaning.
- Valve brush with attached handle
- Ideal for cleaning drums, tanks, buckets
- Polyester bristles, plastic handle
- Available in a range of sizes
All materials of construction are produced from base resins. A model is available without handle for use with STA series handles.
Certifications: BPA-free and FDA approved.
Frequently Bought Together