J-Zone Floor Drain Brush and Handles, Micronova
Supplier: Micronova
Certificates
About this item
A stiff-bristle brush head and threaded plastic handle, specifically designed for cleaning floor drains. The brush head features black bristles for color-coding and the handle is printed with 'For Floor Drain Use Only' to keep separate from other cleaning brushes.
- Stiff bristle, replaceable brush head
- Handle is printed with 'For Floor Drain Use Only'
- Clear distinction from other cleaning brushes
- 4" (10 cm) diameter brush head
- Long, 36" (91 cm) handle to easily reach floor
The Micronova J-Zone Floor Drain Brush head features a plastic threaded end for easy replacement with the Floor Drain Handle.
Ordering information: Brush and handle must be ordered separately.
Frequently Bought Together