Top Buret, digital burettes, Eppendorf
   Sustainable Options Available
Supplier:   Eppendorf
Top Buret, digital burettes, Eppendorf
Supplier:  Eppendorf
   Sustainable Options Available
Certificates
Top Buret, digital burettes, Eppendorf
2 Options Available Below
Product Details & Documents
Continuous dispensing from 0.01 to 999.9 mL by rotating the wheels: In increments of 2.5 ml per complete rotation for the Top Buret.
  • M model and 5 mL per rotation for the Top Buret H model
  • Safety valve for priming without loss of reagent
  • Discharge tube can be rotated through 360° and adjusted vertically (10 - 200 mm) and horizontally (142 - 220 mm)
  • Connection for dry tube - available as option
  • Power supplied by long-life batteries with charging display
For safe, simple, accurate and continuous titration.
 : Supplied with safety valve, telescopic filling tube (210 - 370 mm), adjustable discharge tube, three adapters for 32/38/40 mm diameter bottle threads, two micro batteries and a certificate of quality.
Specifications for Product Family
Specifications
  • Catalog No:
  • CA89133-972
  • CA89133-974
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
Product Information
  • Désignation
    Volume
    Subdivisions
    Availability
    Price
  • Top Buret M, 25 ml
    0.01 - 999.9 ml
    2.5 ml per turn
    Log in for availability
    Log in for availability
    Each
    Each - $2,640.00
    $2,640.00
    Each
       $2,640.00
    Supplier Number: 4965000017
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    CA89133-972
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Top Buret H, 50 ml
    0.01 - 999.9 ml
    5 ml per turn
    Log in for availability
    Log in for availability
    Each
    Each - $2,640.00
    $2,640.00
    Each
       $2,640.00
    Supplier Number: 4965000025
    Specifications:
    More
    Specifications
    Cat. No.
    CA89133-974
    Environmentally Preferable
    Environmental certification