ContactUsLinkComponent
Scoop, Stainless Steel, Bürkle
Catalogue # 10217-178
Fournisseur:  BURKLE INC
undefined
Scoop, Stainless Steel, Bürkle
Catalogue # 10217-178
Fournisseur:  BURKLE INC
Supplier Number:  5354-1000
Spécifications
  • Capacity:
    1000 ml
  • Length:
    115 cm
  • Cat. no.:
    10217-178
  • Supplier No.:
    5354-1000
Spécifications
À propos de cet article
The stainless steel scoop has no openings, undercuts or crevices in which residues could collect
Pore-free finishing produced by high-quality laser welding seams. The stainless steel scoop can be flame-sterilised and can therefore be made completely germ-free.