SiloDrill Sampler, Bürkle | Avantor
SiloDrill Sampler, Bürkle
Supplier:   BURKLE INC
SiloDrill Sampler, Bürkle
Supplier:  BURKLE INC
Certificates
SiloDrill Sampler, Bürkle
SiloDrill Sampler, Bürkle
2 Options Available Below
Product Details & Documents
Representative samples from all layers of the silo can be taken with the SiloDrill.
  • Made of stainless steel V2A (1.4301) or aluminium
  • Standard length 150 cm, extendable in increments of 100 cm, max. 350 cm
  • Sprung inner tube prevents jamming of coarse grains
  • Scraper edge for cohesive substances
  • Sturdy, removable T-handle
  • Drill tip with drilling blades for easy boring
  • Easy removal of the sample chamber, simple cleaning
The SiloDrill fits through all standard openings with its diameter of 90 mm (3.5").
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Capacité
    Longueur
    Ø
    Availability
    Price
  • Sampler, SiloDrill
    Aluminium
    400 ml (13.5 oz.)
    150 cm (59")
    40 mm (1.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,764.37
    $2,764.37
    Each
       $2,764.37
    Supplier Number: 5301-1000
  • Sampler, SiloDrill
    Stainless steel V2A
    400 ml (13.5 oz.)
    150 cm (59")
    40 mm (1.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13,752.27
    $13,752.27
    Each
       $13,752.27
    Supplier Number: 5301-2000
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • SiloDrill extension V2A, 100 cm
    SiloDrill extension V2A, 100 cm
    10216-970
    Log in pour la disponibilité
    Each - $555.20